accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 365-34-4

IUPAC Name: 1-phenyl-2-(trifluoromethyl)hydrazine
Synonyms: 1-phenyl-2-(trifluoromethyl)hydrazine
Molecular Weight (g/mol): 176.14
Molecular Formula: C7H7F3N2
InChi Key: LJORWWOXCXNLOT-UHFFFAOYSA-N
SMILES: FC(F)(F)NNC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results