accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)NH3OO

CAS RN 3653-34-7

IUPAC Name: (3S)-3-azaniumyl-5-methylhexanoate
Synonyms: (3S)-3-ammonio-5-methylhexanoate
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: MLYMSIKVLAPCAK-LURJTMIESA-N
SMILES: CC(C)C[C@H]([NH3+])CC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results