accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNNNSAg(E/Z)(E/Z)

CAS RN 36539-86-3

IUPAC Name: silver(1+) [N'-(phenylamino)-N-(phenylimino)carbamimidoyl]sulfanide
Synonyms: silver(1+) [N'-(phenylamino)-N-(phenylimino)carbamimidoyl]sulfanide
Molecular Weight (g/mol): 363.19
Molecular Formula: C13H11AgN4S
InChi Key: RTNZRONZKMOMBV-UHFFFAOYSA-M
SMILES: [Ag+].[S-]C(=NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results