accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COCH3OCH3

CAS RN 365564-07-4

IUPAC Name: 2-(3,5-dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,5-dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 264.13
Molecular Formula: C14H21BO4
InChi Key: CZYHRTIJLUONKY-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results