accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 366-29-0

IUPAC Name: N4,N4,N4',N4'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N,N,N',N'-tetramethylbenzidine
Molecular Weight (g/mol): 240.35
Molecular Formula: C16H20N2
InChi Key: YRNWIFYIFSBPAU-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)