accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 366-99-4

IUPAC Name: 3-fluoro-4-methoxyaniline
Synonyms: 3-fluoro-4-methoxyaniline
Molecular Weight (g/mol): 141.15
Molecular Formula: C7H8FNO
InChi Key: LJWAPDSCYTZUJU-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C=C1F
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)