accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 366017-09-6

IUPAC Name: 1-{4-[4-({2-[(1E)-2-[4-(trifluoromethyl)phenyl]ethenyl]-1,3-oxazol-4-yl}methoxy)phenyl]butyl}-1H-1,2,3-triazole
Synonyms: mubritinib
Molecular Weight (g/mol): 468.48
Molecular Formula: C25H23F3N4O2
InChi Key: ZTFBIUXIQYRUNT-MDWZMJQESA-N
SMILES: FC(F)(F)C1=CC=C(\C=C\C2=NC(COC3=CC=C(CCCCN4C=CN=N4)C=C3)=CO2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results