CAS RN 36603-80-2
IUPAC Name:
sodium tricyanomethanide
Synonyms:
sodium tricyanomethanide
Molecular Weight (g/mol):
113.06
Molecular Formula:
C4N3Na
InChi Key:
LOYHZHALCDOXIK-UHFFFAOYSA-N
SMILES:
[Na+].N#C[C-](C#N)C#N
1
–
1
of
1
results
1
Sodium Tricyanomethanide 98.0+%, TCI America™
CAS: 36603-80-2 Molecular Formula: C4N3Na Molecular Weight (g/mol): 113.055 InChI Key: LOYHZHALCDOXIK-UHFFFAOYSA-N Synonym: Tricyanomethanide Sodium Salt PubChem CID: 24840070 IUPAC Name: sodium;methanetricarbonitrile SMILES: C(#N)[C-](C#N)C#N.[Na+]