accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOO

CAS RN 3669-41-8

IUPAC Name: 1-(2,4-dihydroxyphenyl)-2-phenylethan-1-one
Synonyms: 1-(2,4-dihydroxyphenyl)-2-phenylethanone
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: VFQKAJVKZKHVPD-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(=O)CC2=CC=CC=C2)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results