accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 367-22-6

IUPAC Name: 4-chloro-3-fluoroaniline
Synonyms: 4-chloro-3-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: ACMJJQYSPUPMPN-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(F)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)