accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FF

CAS RN 367-25-9

IUPAC Name: 2,4-difluoroaniline
Synonyms: 2,4-difluoroaniline
Molecular Weight (g/mol): 129.11
Molecular Formula: C6H5F2N
InChi Key: CEPCPXLLFXPZGW-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=C1F
Boiling Point 
  • (3)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)