accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFFFFF

CAS RN 367-71-5

IUPAC Name: 2,4-bis(trifluoromethyl)aniline
Synonyms: 2,4-bis(trifluoromethyl)aniline
Molecular Weight (g/mol): 229.13
Molecular Formula: C8H5F6N
InChi Key: UIWVOUBVGBJRNP-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results