accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FNOONOO

CAS RN 367-81-7

IUPAC Name: 5-fluoro-2,4-dinitroaniline
Synonyms: 5-fluoro-2,4-dinitroaniline
Molecular Weight (g/mol): 201.11
Molecular Formula: C6H4FN3O4
InChi Key: RAGRTYREMCPEIV-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results