accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3672-47-7

IUPAC Name: 3-(4-methoxyphenyl)-3-oxopropanenitrile
Synonyms: 3-(4-methoxyphenyl)-3-oxopropanenitrile
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: IKEPUFCALLUUBC-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CC#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)