accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36725-28-7

IUPAC Name: 6-(4-aminophenyl)-5-methyl-2,3,4,5-tetrahydropyridazin-3-one
Synonyms: 6-(4-aminophenyl)-5-methyl-4,5-dihydro-2H-pyridazin-3-one
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: GDMRFHZLKNYRRO-UHFFFAOYNA-N
SMILES: CC1CC(=O)NN=C1C1=CC=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results