accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36727-29-4

IUPAC Name: 3,5,5-trimethylhexanoyl chloride
Synonyms: 3,5,5-trimethylhexanoyl chloride
Molecular Weight (g/mol): 176.68
Molecular Formula: C9H17ClO
InChi Key: GEKPNPPFAYJZRD-UHFFFAOYNA-N
SMILES: CC(CC(Cl)=O)CC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results