accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHN

CAS RN 36729-27-8

IUPAC Name: 2,3,4,9-tetrahydro-1H-carbazole-6-carboxylate
Synonyms: 6,7,8,9-tetrahydro-5H-carbazole-3-carboxylate
Molecular Weight (g/mol): 214.25
Molecular Formula: C13H12NO2
InChi Key: OWQQDAGRTDUORV-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2NC3=C(CCCC3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results