accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNHONONOClCl

CAS RN 36734-19-7

IUPAC Name: 3-(3,5-dichlorophenyl)-2,4-dioxo-N-(propan-2-yl)imidazolidine-1-carboxamide
Synonyms: iprodione rovral rovral (iprodione)
Molecular Weight (g/mol): 330.17
Molecular Formula: C13H13Cl2N3O3
InChi Key: ONUFESLQCSAYKA-UHFFFAOYSA-N
SMILES: CC(C)NC(=O)N1CC(=O)N(C1=O)C1=CC(Cl)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results