accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3FFFSOOO

CAS RN 367522-96-1

IUPAC Name: 1-butyl-1-methylpyrrolidin-1-ium trifluoromethanesulfonate
Synonyms: 1-butyl-1-methylpyrrolidin-1-ium triflate
Molecular Weight (g/mol): 291.33
Molecular Formula: C10H20F3NO3S
InChi Key: WZJDNKTZWIOOJE-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CCCC[N+]1(C)CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results