accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36760-20-0

IUPAC Name: 3,5-dichloro-2H-indazole
Synonyms: 3,5-dichloro-2H-indazole
Molecular Weight (g/mol): 187.02
Molecular Formula: C7H4Cl2N2
InChi Key: QQZISIWJMCCCNH-UHFFFAOYSA-N
SMILES: ClC1=C2C=C(Cl)C=CC2=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results