accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClClN

CAS RN 36764-94-0

IUPAC Name: 2-amino-3,5-dichlorobenzonitrile
Synonyms: 2-amino-3,5-dichlorobenzonitrile
Molecular Weight (g/mol): 187.02
Molecular Formula: C7H4Cl2N2
InChi Key: ZHKNDJRPOVUPMT-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Cl)C=C1C#N
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)