accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2FFF

CAS RN 368-53-6

IUPAC Name: 5-(trifluoromethyl)benzene-1,3-diamine
Synonyms: 5-(trifluoromethyl)benzene-1,3-diamine
Molecular Weight (g/mol): 176.14
Molecular Formula: C7H7F3N2
InChi Key: KZSXRDLXTFEHJM-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(N)=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)