accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3680-71-5

IUPAC Name: 1H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one
Synonyms: 1H,7H-pyrrolo[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 135.13
Molecular Formula: C6H5N3O
InChi Key: FBMZEITWVNHWJW-UHFFFAOYSA-N
SMILES: O=C1N=CNC2=C1C=CN2
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)