accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36809-26-4

IUPAC Name: 4-bromo-N,N-diphenylaniline
Synonyms: 4-bromo-N,N-diphenylaniline
Molecular Weight (g/mol): 324.22
Molecular Formula: C18H14BrN
InChi Key: SQTLUXJWUCHKMT-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)