accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36821-26-8

IUPAC Name: ethyl 2-oxopyrrolidine-3-carboxylate
Synonyms: ethyl 2-oxopyrrolidine-3-carboxylate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: FHHMWJMUMOYGQW-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCNC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results