accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOHAsOHONHNOSOOOSOOO(E)

CAS RN 3688-92-4

IUPAC Name: disodium (4E)-4-[2-(2-arsonophenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: thorin (chemistry)
Molecular Weight (g/mol): 576.29
Molecular Formula: C16H11AsN2Na2O10S2
InChi Key: ZQMPNLOZJUWHSA-WTBAQZMLSA-L
SMILES: [Na+].[Na+].O[As](O)(=O)C1=CC=CC=C1N\N=C1\C(=O)C(=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)