accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36881-42-2

IUPAC Name: 1-(bromomethyl)-4-phenoxybenzene
Synonyms: 1-(bromomethyl)-4-phenoxybenzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: CPIGBCFBFZSCQI-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(OC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)