accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 368869-86-7

IUPAC Name: 1-(4-iodophenyl)-1H-pyrazole
Synonyms: 1-(4-iodophenyl)pyrazole
Molecular Weight (g/mol): 270.07
Molecular Formula: C9H7IN2
InChi Key: ADIQBEMLKKKISJ-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results