accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3OPSOCH3OCH3

CAS RN 3689-24-5

IUPAC Name: O,O-diethyl {[diethoxy(sulfanylidene)-λ⁵-phosphanyl]oxy}phosphonothioate
Synonyms: dithione caswell no. 837 fulex Sulpho-TEPP rcra waste number P109 tetraethyl ester dithiodiphosphoric acid tetraethyldithiopyrophosphate lethalaire G-57 ethyl thiopyrophosphate ((eto)2(PS)2O) Sulfo-TEPP
Molecular Weight (g/mol): 322.31
Molecular Formula: C8H20O5P2S2
InChi Key: XIUROWKZWPIAIB-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OP(=S)(OCC)OCC
Element 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)