accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3693-53-6

IUPAC Name: ethyl N-{[(ethoxycarbonyl)amino]methyl}carbamate
Synonyms: ethyl N-{[(ethoxycarbonyl)amino]methyl}carbamate
Molecular Weight (g/mol): 190.2
Molecular Formula: C7H14N2O4
InChi Key: QSPOJWMZYOMAEI-UHFFFAOYSA-N
SMILES: CCOC(=O)NCNC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)