accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36930-63-9

IUPAC Name: 5-{[2-(2-iodoacetamido)ethyl]amino}naphthalene-1-sulfonic acid
Synonyms: iaedans 1,5-I-aedans I-aed
Molecular Weight (g/mol): 434.25
Molecular Formula: C14H15IN2O4S
InChi Key: ZMERMCRYYFRELX-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=C2C=CC=C(NCCNC(=O)CI)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results