accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 369405-27-6

IUPAC Name: tetrafluoroboranuide; {[bis(propan-2-yl)amino]methylidene}bis(propan-2-yl)azanium
Synonyms: [(diisopropylamino)methylidene]diisopropylazanium tetrafluoroborate
Molecular Weight (g/mol): 300.19
Molecular Formula: C13H29BF4N2
InChi Key: PEKVUVXZBLFOHV-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC(C)N(C=[N+](C(C)C)C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results