accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOH2NOHO

CAS RN 3695-73-6

IUPAC Name: 2-(2-aminoacetamido)propanoic acid
Synonyms: 2-(2-aminoacetamido)propanoic acid
Molecular Weight (g/mol): 146.15
Molecular Formula: C5H10N2O3
InChi Key: VPZXBVLAVMBEQI-UHFFFAOYNA-N
SMILES: CC(NC(=O)CN)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)