accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3695-77-0

IUPAC Name: triphenylmethanethiol
Synonyms: triphenylmethanethiol
Molecular Weight (g/mol): 276.4
Molecular Formula: C19H16S
InChi Key: JQZIKLPHXXBMCA-UHFFFAOYSA-N
SMILES: SC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (6)
Quantity 
  • (3)
  • (4)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (6)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)