accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3696-66-0

IUPAC Name: 1-(isothiocyanatomethyl)-3-methylbenzene
Synonyms: 1-(isothiocyanatomethyl)-3-methylbenzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: NUXFAFWRXGFTQJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CN=C=S)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results