accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3NH2

CAS RN 36966-04-8

IUPAC Name: N-ethylnaphthalen-1-aminium bromide
Synonyms: N-ethylnaphthalen-1-aminium bromide
Molecular Weight (g/mol): 252.16
Molecular Formula: C12H14BrN
InChi Key: SBGSNWFFTCHTQG-UHFFFAOYSA-N
SMILES: [Br-].CC[NH2+]C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results