accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClNH2NHClNNH2NNNH2NNH2HNCl(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 3697-42-5

IUPAC Name: dihydrogen N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide dichloride
Synonyms: chlorhexidine dihydrochloride
Molecular Weight (g/mol): 578.37
Molecular Formula: C22H32Cl4N10
InChi Key: WJLVQTJZDCGNJN-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)