accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3698-95-1

IUPAC Name: 1-(methylsulfanyl)octane
Synonyms: 1-(methylsulfanyl)octane
Molecular Weight (g/mol): 160.32
Molecular Formula: C9H20S
InChi Key: AHCJTMBRROLNHV-UHFFFAOYSA-N
SMILES: CCCCCCCCSC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results