accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOS

CAS RN 36983-36-5

IUPAC Name: ethyl 2,4-dioxo-4-(thiophen-2-yl)butanoate
Synonyms: ethyl 2,4-dioxo-4-(thiophen-2-yl)butanoate
Molecular Weight (g/mol): 226.25
Molecular Formula: C10H10O4S
InChi Key: GCCFYXYKJZUHMI-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CC(=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results