accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH3CNH3CCH3CH3

CAS RN 3700-67-2

IUPAC Name: dimethyldioctadecylazanium bromide
Synonyms: dimethyldioctadecylazanium bromide
Molecular Weight (g/mol): 630.97
Molecular Formula: C38H80BrN
InChi Key: PSLWZOIUBRXAQW-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)