accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3O(S)P(S)PO

CAS RN 37002-48-5

IUPAC Name: {[(4S,5S)-5-[(diphenylphosphanyl)methyl]-2,2-dimethyl-1,3-dioxolan-4-yl]methyl}diphenylphosphane
Synonyms: {[(4S,5S)-5-[(diphenylphosphanyl)methyl]-2,2-dimethyl-1,3-dioxolan-4-yl]methyl}diphenylphosphane
Molecular Weight (g/mol): 498.54
Molecular Formula: C31H32O2P2
InChi Key: VCHDBLPQYJAQSQ-LOYHVIPDSA-N
SMILES: CC1(C)O[C@H](CP(C2=CC=CC=C2)C2=CC=CC=C2)[C@@H](CP(C2=CC=CC=C2)C2=CC=CC=C2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results