accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 370069-31-1

IUPAC Name: tert-butyl 2-(aminomethyl)piperidine-1-carboxylate
Synonyms: tert-butyl 2-(aminomethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: PTVRCUVHYMGECC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCCC1CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results