CAS RN 3704-28-7
IUPAC Name:
methyl 2-(methylsulfanyl)benzoate
Synonyms:
methyl 2-(methylsulfanyl)benzoate
Molecular Weight (g/mol):
182.24
Molecular Formula:
C9H10O2S
InChi Key:
CPQDZXPLQXZJGF-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC=C1SC
1
–
1
of
1
results
1
Promotions Available
Methyl 2-(methylthio)benzoate, 98%
Methyl 2-(methylthio)benzoate, 98%
CAS: 3704-28-7 Molecular Formula: C9H10O2S Molecular Weight (g/mol): 182.237 MDL Number: MFCD00068039 InChI Key: CPQDZXPLQXZJGF-UHFFFAOYSA-N PubChem CID: 279586 IUPAC Name: methyl 2-methylsulfanylbenzoate SMILES: COC(=O)C1=CC=CC=C1SC