accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOHOSOOO

CAS RN 37067-27-9

IUPAC Name: potassium 4-hydroxyphenyl sulfate
Synonyms: potassium 4-hydroxyphenyl sulfate
Molecular Weight (g/mol): 228.26
Molecular Formula: C6H5KO5S
InChi Key: KYQHUXZXYUBUPX-UHFFFAOYSA-M
SMILES: [K+].OC1=CC=C(OS([O-])(=O)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results