accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37073-15-7

IUPAC Name: 1-methanesulfonyl-1H-1,2,3-benzotriazole
Synonyms: 1-methanesulfonyl-1,2,3-benzotriazole
Molecular Weight (g/mol): 197.21
Molecular Formula: C7H7N3O2S
InChi Key: OMMZSDGGKVRGGP-UHFFFAOYSA-N
SMILES: CS(=O)(=O)N1N=NC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results