accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOO

CAS RN 3708-04-1

IUPAC Name: 2-methanesulfonyl-1-phenylethan-1-one
Synonyms: 2-methanesulfonyl-1-phenylethanone
Molecular Weight (g/mol): 198.24
Molecular Formula: C9H10O3S
InChi Key: CAUSPZIZBLGLKW-UHFFFAOYSA-N
SMILES: CS(=O)(=O)CC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results