accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNHNNClClClClClClNHCl

CAS RN 37083-37-7

IUPAC Name: 2,7,12,17-tetrakis(2,6-dichlorophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Synonyms: 2,7,12,17-tetrakis(2,6-dichlorophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular Weight (g/mol): 890.29
Molecular Formula: C44H22Cl8N4
InChi Key: LOKNIYQQLQPXMC-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=C(Cl)C=CC=C1Cl)C1=C(Cl)C=CC=C1Cl)C1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results