CAS RN 3709-08-8
CAS RN 3709-08-8
IUPAC Name:
3,3-dimethyloxolan-2-one
Synonyms:
3,3-dimethyloxolan-2-one
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
UPVAIJPDWVTFKT-UHFFFAOYSA-N
SMILES:
CC1(C)CCOC1=O
1
–
1
of
1
results
1
alpha,alpha-Dimethyl-gamma-butyrolactone 97.0+%, TCI America™
CAS: 3709-08-8 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.14 MDL Number: MFCD03844788 InChI Key: UPVAIJPDWVTFKT-UHFFFAOYSA-N Synonym: Dihydro-3,3-dimethyl-2(3H)-furanone PubChem CID: 94746 IUPAC Name: 3,3-dimethyloxolan-2-one SMILES: CC1(CCOC1=O)C