CAS RN 3709-18-0
IUPAC Name:
2,2,5-trimethyl-1,3-dioxane-4,6-dione
Synonyms:
2,2,5-trimethyl-1,3-dioxane-4,6-dione
Molecular Weight (g/mol):
158.15
Molecular Formula:
C7H10O4
InChi Key:
KJMCAXYHYPDRAV-UHFFFAOYSA-N
SMILES:
CC1C(=O)OC(C)(C)OC1=O
1
–
1
of
1
results
1
Sigma Aldrich 2,2,5-Trimethyl-1,3-dioxane-4,6-dione
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.