accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSF

CAS RN 371-15-3

IUPAC Name: 1-fluoro-4-(methylsulfanyl)benzene
Synonyms: 1-fluoro-4-(methylsulfanyl)benzene
Molecular Weight (g/mol): 142.19
Molecular Formula: C7H7FS
InChi Key: XFUMHENRNCUHOH-UHFFFAOYSA-N
SMILES: CSC1=CC=C(F)C=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)